Learn More
Sino Biological Recombinant Human CDCP1 Protein (Secreted Form, mFc Tag), HPLC-verified
Supplier:  Sino Biological 13262H05H50
Encompass_Preferred
A DNA sequence encoding the human CDCP1 (Q9H5V8-3) (Met1-Glu343) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8472