Learn More
Sino Biological Recombinant Human LCAT Protein (His Tag)
Supplier:  Sino Biological 13271H08H100
Encompass_Preferred
A DNA sequence encoding the human LCAT (NP_000220.1) (Met1-Glu440) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8475