Learn More
Sino Biological Recombinant Human CCDC47 Protein (His Tag)
Supplier:  Sino Biological 13274H08H100
Encompass_Preferred
A DNA sequence encoding the human CCDC47 (Q96A33-1) (Met1-Ser135) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8477