Learn More
Sino Biological Recombinant Human alpha amylase/AMY2A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13277H08H100
Encompass_Preferred
A DNA sequence encoding the human AMY2A (P04746) (Met1-Leu511) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8482