Learn More
Sino Biological Recombinant Human ORM2 Protein (His Tag)
Supplier:  Sino Biological 13299H08H100
Encompass_Preferred
A DNA sequence encoding the human ORM2 (P19652) (Met1-Ser201) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8488