Learn More
Sino Biological Recombinant Human TMEM27 Protein (hFc Tag)
Supplier:  Sino Biological 13301H02H100
Encompass_Preferred
A DNA sequence encoding the human TMEM27 (Q9HBJ8) (Met1-Pro141) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8490