Learn More
Sino Biological Recombinant Human TMEM27 Protein (His Tag)
Supplier:  Sino Biological 13301H08H100
Encompass_Preferred
A DNA sequence encoding the human TMEM27 (NP_065716.1) (Met1-Pro141) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8492