Learn More
Sino Biological Recombinant Human TMED1 Protein (His Tag)
Supplier:  Sino Biological 13302H08H100
Encompass_Preferred
A DNA sequence encoding the human TMED1 (Q13445) (Met1-Asn194) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8496