Learn More
Sino Biological Recombinant Human EPCR Protein (His Tag), HPLC verified
Supplier:  Sino Biological 13320H08H50
Encompass_Preferred
A DNA sequence encoding the extracellular domain of human PROCR (Q9UNN8) (Met 1-Thr 209), was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8503