Learn More
Sino Biological Recombinant Human ERP72 Protein (hFc Tag)
Supplier:  Sino Biological 13333H02H100
Encompass_Preferred
A DNA sequence encoding the human PDIA4 (P13667) (Met1-Thr641) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8512