Learn More
Sino Biological Recombinant Human CLM-9 Protein
Supplier:  Sino Biological 13365HCCH100
Encompass_Preferred
A DNA sequence encoding the human CD300LG (AAH25395.1)(Met1-Arg247) was expressed with six amino acids (LEVLFQ) at the C-terminus was expressed and purified.
Catalog No. 50-161-8528