Learn More
Sino Biological Recombinant Human TEM8/ANTXR1 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 13367H02H50
Encompass_Preferred
A DNA sequence encoding the human ANTXR1 (NP_115584.1) (Met1-Ser321) was expressed, fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8533