Learn More
Sino Biological Recombinant Human LRG1 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13371H08H50
Encompass_Preferred
A DNA sequence encoding the human LRG1 (AAH34389.1)(Met1-Gln347) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-8543