Learn More
Sino Biological Recombinant Human LRG1 Protein, HPLC-verified
Supplier:  Sino Biological 13371HCCH50
Encompass_Preferred
A DNA sequence encoding the human LRG1 (AAH34389.1) (Met1-Gln347) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-161-8545