Learn More
eMolecules 133745-75-2 | N-Fluorobenzenesulfonimide | Oakwood Chemicals | MFCD00144885 | 315.330 | C12H10FNO4S2 | 98.000 | FN(S(=O)(=O)c1ccccc1)S(=O)(=O)c1ccccc1 | 25g | 480105677
Shop All eMolecules Inc. Products