Learn More
Sino Biological Recombinant Human FSTL5 Protein (hFc Tag)
Supplier:  Sino Biological 13408H02H100
Encompass_Preferred
A DNA sequence encoding the human FSTL5 (Q8N475-1)(Met1-Ala847) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8550