Learn More
Sino Biological Recombinant Human FSTL5 Protein (His Tag)
Supplier:  Sino Biological 13408H08H100
Encompass_Preferred
A DNA sequence encoding the human FSTL5 (Q8N475) (Gln32-Ala847) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8552