Learn More
Sino Biological Recombinant Human MMP-26 Protein
Supplier:  Sino Biological 13415HNAE20
Encompass_Preferred
A DNA sequence encoding the human MMP26 (NP_068573.2) (Thr90-Pro261) was expressed and purified with an initial Met.
Catalog No. 50-161-8557