Learn More
Sino Biological Recombinant Human SCGB3A1 Protein (mFc Tag)
Supplier:  Sino Biological 13419H05H100
Encompass_Preferred
A DNA sequence encoding the human SCGB3A1 (NP_443095.2) (Met1-Gly104) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8560