Learn More
Sino Biological Recombinant Human UNC5A Protein (ECD, His Tag)
Supplier:  Sino Biological 13424H08H100
Encompass_Preferred
A DNA sequence encoding the human UNC5A (NP_588610.2) (Met1-Tyr306) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8566