Learn More
Sino Biological Recombinant Human BTN3A3 Protein (mFc Tag)
Supplier:  Sino Biological 13432H05H100
Encompass_Preferred
A DNA sequence encoding the human BTN3A3 (O00478-1) (Met1-Trp248) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8568