Learn More
Sino Biological Recombinant Human BTN3A3 Protein (His Tag)
Supplier:  Sino Biological 13432H08H100
Encompass_Preferred
A DNA sequence encoding the human BTN3A3(O00478-1) (Met1-Trp248) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-8570