Learn More
Sino Biological Recombinant Human SMR3B Protein (mFc Tag)
Supplier:  Sino Biological 13442H05H100
Encompass_Preferred
A DNA sequence encoding the human SMR3B (NP_006676.1) (Met1-Pro79) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8578