Learn More
Sino Biological Recombinant Human SMR3B Protein (His Tag)
Supplier:  Sino Biological 13442H08H100
Encompass_Preferred
A DNA sequence encoding the human SMR3B (NP_006676.1) (Met1-Pro79) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-161-8580