Learn More
Sino Biological Recombinant Human JTB Protein (mFc Tag)
Supplier:  Sino Biological 13447H05H100
Encompass_Preferred
A DNA sequence encoding the human JTB (O76095-1) (Met1-Leu105) was fused with Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8582