Learn More
Sino Biological Recombinant Human ZG16 Protein (mFc Tag)
Supplier:  Sino Biological 13450H05H100
Encompass_Preferred
A DNA sequence encoding the human ZG16 (O60844.1) (Met1-Cys167) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8584