Learn More
Sino Biological Recombinant Human TMP21 Protein (mFc Tag)
Supplier:  Sino Biological 13462H05H100
Encompass_Preferred
A DNA sequence encoding the human TMED10 (NP_006818.3) (Met1-Arg185) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8590