Learn More
Sino Biological Recombinant Human Ribonuclease A Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13468H08H100
Encompass_Preferred
A DNA sequence encoding the human RNASE1 (P07998) (Met1-Thr156) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8592