Learn More
Sino Biological Recombinant Human SF20 Protein (mFc Tag)
Supplier:  Sino Biological 13469H05H50
Encompass_Preferred
A DNA sequence encoding the human C19orf10 (NP_061980.1) (Met1-Leu173) was expressed with the Fc region of mouse IgG1 at the C-terminus.
Catalog No. 50-161-8595