Learn More
Sino Biological Recombinant Human VISTA/B7-H5 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 13482H02H100
Encompass_Preferred
A DNA sequence encoding the human C10orf54 (AAH20568.1) (Met1-Ala194) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8600