Learn More
Sino Biological Recombinant Human Calumenin Protein (His Tag)
Supplier:  Sino Biological 13486H08H100
Encompass_Preferred
A DNA sequence encoding the human CALU (O43852-1) (Met1-Arg311) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8605