Learn More
Sino Biological Recombinant Human GPR37 Protein (His Tag)
Supplier:  Sino Biological 13496H08H100
Encompass_Preferred
A DNA sequence encoding the human GPR37 (NP_005293.1) the first extracellular domaqin (Met 1-Met 265) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8613