Learn More
Sino Biological NRG1 Alpha Protein, Human, Recombinant (ECD, Fc Tag)
Supplier:  Sino Biological 13499H02H100
Encompass_Preferred
A DNA sequence encoding the human NRG1 isoform alpha (Q02297-1) extracellular domain (Ser 20-Lys 241) was fused with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8619