Learn More
Sino Biological NRG1 Alpha Protein, Human, Recombinant (ECD, His Tag)
Supplier:  Sino Biological 13499H08H100
Encompass_Preferred
A DNA sequence encoding the human NRG1 isoform alpha (Q02297-1) extracellular domain (Ser 20-Lys 241) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8621