Learn More
Bio Rad Laboratories 1,350 psi Rupture Disks
Catalog No. NC1489157
Encompass
Pkg of 100, bursts at 1,350 psi in a partial vacuum, for use with the PDS-1000/He biolistic system
Catalog No. NC1489157 Supplier Bio Rad Laboratories Supplier No. 1652330
Pkg of 100, bursts at 1,350 psi in a partial vacuum, for use with the PDS-1000/He biolistic system