Learn More
Sino Biological Recombinant Human SCN3B Protein (His Tag)
Supplier:  Sino Biological 13500H08H100
Encompass_Preferred
A DNA sequence encoding the mature form of human SCN3B (Q9NY72) extracellular domain (Met1-Glu159) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8626