Learn More
Sino Biological Recombinant Human TXNL4B Protein (His Tag)
Supplier:  Sino Biological 13501H07E100
Encompass_Preferred
A DNA sequence encoding the human TXNL4B (Q9NX01) (Met 1-Ile 149) was expressed, with a polyhistide tag at the N-terminus.
Catalog No. 50-161-8628