Learn More
Sino Biological Recombinant Human IZUMO1 Protein (His Tag)
Supplier:  Sino Biological 13520H08H100
Encompass_Preferred
A DNA sequence encoding the human IZUMO1 (AAH34769.1) (Met1-Arg292) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8650