Learn More
Sino Biological Recombinant Human Reticulocalbin 3/RCN3 Protein (His Tag)
Supplier:  Sino Biological 13533H08H100
Encompass_Preferred
A DNA sequence encoding the human RCN3 (Q96D15) (Met1-His324) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8654