Learn More
Sino Biological Recombinant Human PRELP Protein (mFc Tag)
Supplier:  Sino Biological 13538H04H100
Encompass_Preferred
A DNA sequence encoding the human PRELP (NP_002716.1) (Gln21-Ile382) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-161-8658