Learn More
Sino Biological Recombinant Human PRELP Protein
Supplier:  Sino Biological 13538HNCH100
Encompass_Preferred
A DNA sequence encoding the human PRELP (NP_002716.1) (Gln21-Ile382) was expressed with two additional amino acids (Gly & Pro) at the N-terminus.
Catalog No. 50-161-8660