Learn More
Sino Biological Recombinant Human Calreticulin/CALR Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13539H08H50
Encompass_Preferred
A DNA sequence encoding the human CALR (P27797) (Met1-Ala413) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8665