Learn More
Sino Biological Recombinant Human DPEP1 Protein (His Tag)
Supplier:  Sino Biological 13543H08H50
Encompass_Preferred
A DNA sequence encoding the human DPEP1 (P16444) (Met1-Ser385) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8669