Learn More
Sino Biological Recombinant Human ADAMTSL1 Protein (His Tag)
Supplier:  Sino Biological 13550H08B20
Encompass_Preferred
A DNA sequence encoding the human ADAMTSL1 (Met 1-His439) (Q8N6G6-2) was expressed, with a C-terminal polyhistidine tag.
Catalog No. 50-161-8679