Learn More
Sino Biological Recombinant Human SMPDL3A Protein (His Tag)
Supplier:  Sino Biological 13559H08H100
Encompass_Preferred
A DNA sequence encoding the human SMPDL3A (Q92484) (Met1-Tyr453) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8688