Learn More
Sino Biological Recombinant Human beta Amylase/AMY2B Protein (hFc Tag)
Supplier:  Sino Biological 13575H02H100
Encompass_Preferred
A DNA sequence encoding the human AMY2B (NP_066188.1) (Met1-Leu511) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8694