Learn More
Electron Microscopy Sciences Flim Sleeve 135 mm/20 exp with Clear open flap
SDP
Catalog No. 5028009
Encompass_Preferred
Polyguard Polypropylene film sleeves are the finest archival quality material, and are guaranteed to give you ultimate protection for your most important film negatives. Polyguard is 100% acid free, as well as being free from optical distortion. It is resistant to water, grease, oil, alkalies, and organic solvents. Polyguard is durable, yet pliable and gentle to the film and will not become brittle, crack, or abrade the film.
Type: Clear open flap
Catalog No. 50-280-09 Supplier Electron Microscopy Sciences Supplier No. 73930
Polyguard Polypropylene film sleeves are the finest archival quality material, and are guaranteed to give you ultimate protection for your most important film negatives. Polyguard is 100% acid free, as well as being free from optical distortion. It is resistant to water, grease, oil, alkalies, and organic solvents. Polyguard is durable, yet pliable and gentle to the film and will not become brittle, crack, or abrade the film.
Type: Clear open flap