Learn More
Sino Biological Recombinant Human UNC5H2/UNC5B Protein (ECD, hFc Tag)
Supplier:  Sino Biological 13606H02H100
Encompass_Preferred
A DNA sequence encoding the human UNC5B (NP_734465.2) (Met1-Pro363) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-161-8700