Learn More
Sino Biological Recombinant Human Serum Amyloid P Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13610H08H100
Encompass_Preferred
A DNA sequence encoding the human APCS (P02743) (Met1-Val223) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8704