Learn More
Sino Biological Recombinant Human CLPS Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 13631H08B100
Encompass_Preferred
A DNA sequence encoding the human CLPS (P04118) (Met 1-Gln 112) was fused with a polyhistidine tag at the C-terminus.
Catalog No. 50-161-8716